Merck
  • Inosine Monophosphate Dehydrogenase Dependence in a Subset of Small Cell Lung Cancers.
Inosine Monophosphate Dehydrogenase Dependence in a Subset of Small Cell Lung Cancers.
Cell metabolism (2018-07-26)
Fang Huang, Min Ni, Milind D Chalishazar, Kenneth E Huffman, Jiyeon Kim, Ling Cai, Xiaolei Shi, Feng Cai, Lauren G Zacharias, Abbie S Ireland, Kailong Li, Wen Gu, Akash K Kaushik, Xin Liu, Adi F Gazdar, Trudy G Oliver, John D Minna, Zeping Hu, Ralph J DeBerardinis
ABSTRACT
Small cell lung cancer (SCLC) is a rapidly lethal disease with few therapeutic options. We studied metabolic heterogeneity in SCLC to identify subtype-selective vulnerabilities. Metabolomics in SCLC cell lines identified two groups correlating with high or low expression of the Achaete-scute homolog-1 (ASCL1) transcription factor (ASCL1High and ASCL1Low), a lineage oncogene. Guanosine nucleotides were elevated in ASCL1Low cells and tumors from genetically engineered mice. ASCL1Low tumors abundantly express the guanosine biosynthetic enzymes inosine monophosphate dehydrogenase-1 and -2 (IMPDH1 and IMPDH2). These enzymes are transcriptional targets of MYC, which is selectively overexpressed in ASCL1Low SCLC. IMPDH inhibition reduced RNA polymerase I-dependent expression of pre-ribosomal RNA and potently suppressed ASCL1Low cell growth in culture, selectively reduced growth of ASCL1Low xenografts, and combined with chemotherapy to improve survival in genetic mouse models of ASCL1Low/MYCHigh SCLC. The data define an SCLC subtype-selective vulnerability related to dependence on de novo guanosine nucleotide synthesis.
Sigma-Aldrich
Anti-IMPDH1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Mizoribine, ≥98% (TLC)
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture