Merck
  • GLP-1 improves adipose tissue glyoxalase activity and capillarization improving insulin sensitivity in type 2 diabetes.
GLP-1 improves adipose tissue glyoxalase activity and capillarization improving insulin sensitivity in type 2 diabetes.
Pharmacological research (2020-09-18)
Tiago Rodrigues, Patrícia Borges, Laura Mar, Daniela Marques, Miguel Albano, Hans Eickhoff, Catarina Carrêlo, Bruno Almeida, Salomé Pires, Margarida Abrantes, Beatriz Martins, Cristina Uriarte, Filomena Botelho, Pedro Gomes, Sónia Silva, Raquel Seiça, Paulo Matafome
ABSTRACT
Methylglyoxal was shown to impair adipose tissue capillarization and insulin sensitivity in obese models. We hypothesized that glyoxalase-1 (GLO-1) activity could be diminished in the adipose tissue of type 2 diabetic obese patients. Moreover, we assessed whether such activity could be increased by GLP-1-based therapies in order to improve adipose tissue capillarization and insulin sensitivity. GLO-1 activity was assessed in visceral adipose tissue of a cohort of obese patients. The role of GLP-1 in modulating GLO-1 was assessed in type 2 diabetic GK rats submitted to sleeve gastrectomy or Liraglutide treatment, in the adipose tissue angiogenesis assay and in the HUVEC cell line. Glyoxalase-1 activity was decreased in visceral adipose tissue of pre-diabetic and diabetic obese patients, together with other markers of adipose tissue dysfunction and correlated with increased HbA1c levels. Decreased adipose tissue GLO-1 levels in GK rats were increased by sleeve gastrectomy and Liraglutide, being associated with overexpression of angiogenic and vasoactive factors, as well as insulin receptor phosphorylation (Tyr1161). Moreover, GLP-1 increased adipose tissue capillarization and HUVEC proliferation in a glyoxalase-dependent manner. Lower adipose tissue GLO-1 activity was observed in dysmetabolic patients, being a target for GLP-1 in improving adipose tissue capillarization and insulin sensitivity.
Sigma-Aldrich
Glyoxalase I Activity Assay Kit, sufficient for 100 tests (UV)
Pricing and availability is not currently available.
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Pricing and availability is not currently available.