Merck
  • Differential transcriptome and metabolome analysis of Plumbago zeylanica L. reveal putative genes involved in plumbagin biosynthesis.
Differential transcriptome and metabolome analysis of Plumbago zeylanica L. reveal putative genes involved in plumbagin biosynthesis.
Fitoterapia (2020-10-19)
Arati P Vasav, Anupama A Pable, Vitthal T Barvkar
ABSTRACT
Plumbagin is a pharmacologically active naphthoquinone present in the Plumbago zeylanica L. having important medicinal properties. The root of P. zeylanica is rich and primary tissue of the plumbagin biosynthesis and accumulation. The complete biosynthetic pathway of plumbagin in plant is still obscure. The present study attempts to understand the plumbagin biosynthetic pathway with the help of differential transcriptome and metabolome analysis of P. zeylanica leaf and root. The transcriptome data showed co-expression of Aldo-keto reductase (PzAKR), Polyketide cyclase (Pzcyclase) and Cytochrome P450 (PzCYPs) transcripts along with the Polyketide synthase (PzPKS) transcripts. Their higher expression in root as compared to leaf supports their possible involvement in plumbagin biosynthesis. The metabolome data of leaf and root revealed naphthalene derivative isoshinanolone that could be potential precursor of plumbagin. Pathway elucidation and transcriptome data of P. zeylanica, will enable and accelerate research on naphthoquinone biosynthesis in plants.
Product Description
Plumbagin, phyproof® Reference Substance
Pricing and availability is not currently available.
Sigma-Aldrich
Plumbagin from Plumbago indica
Pricing and availability is not currently available.