Merck
  • Temporal Proteomic Analysis of Herpes Simplex Virus 1 Infection Reveals Cell-Surface Remodeling via pUL56-Mediated GOPC Degradation.
Temporal Proteomic Analysis of Herpes Simplex Virus 1 Infection Reveals Cell-Surface Remodeling via pUL56-Mediated GOPC Degradation.
Cell reports (2020-10-08)
Timothy K Soh, Colin T R Davies, Julia Muenzner, Leah M Hunter, Henry G Barrow, Viv Connor, Clément R Bouton, Cameron Smith, Edward Emmott, Robin Antrobus, Stephen C Graham, Michael P Weekes, Colin M Crump
ABSTRACT
Herpesviruses are ubiquitous in the human population and they extensively remodel the cellular environment during infection. Multiplexed quantitative proteomic analysis over the time course of herpes simplex virus 1 (HSV-1) infection was used to characterize changes in the host-cell proteome and the kinetics of viral protein production. Several host-cell proteins are targeted for rapid degradation by HSV-1, including the cellular trafficking factor Golgi-associated PDZ and coiled-coil motif-containing protein (GOPC). We show that the poorly characterized HSV-1 pUL56 directly binds GOPC, stimulating its ubiquitination and proteasomal degradation. Plasma membrane profiling reveals that pUL56 mediates specific changes to the cell-surface proteome of infected cells, including loss of interleukin-18 (IL18) receptor and Toll-like receptor 2 (TLR2), and that cell-surface expression of TLR2 is GOPC dependent. Our study provides significant resources for future investigation of HSV-host interactions and highlights an efficient mechanism whereby a single virus protein targets a cellular trafficking factor to modify the surface of infected cells.
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-GFP, N-terminal antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Millipore
ReBlot Plus Kit
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture