Merck
  • Sanguinarine (pseudochelerythrine) is a potent inhibitor of NF-kappaB activation, IkappaBalpha phosphorylation, and degradation.
Sanguinarine (pseudochelerythrine) is a potent inhibitor of NF-kappaB activation, IkappaBalpha phosphorylation, and degradation.
The Journal of biological chemistry (1997-12-31)
M M Chaturvedi, A Kumar, B G Darnay, G B Chainy, S Agarwal, B B Aggarwal
ABSTRACT
The nuclear factor NF-kappaB is a pleiotropic transcription factor whose activation results in inflammation, viral replication, and growth modulation. Due to its role in pathogenesis, NF-kappaB is considered a key target for drug development. In the present report we show that sanguinarine (a benzophenanthridine alkaloid), a known anti-inflammatory agent, is a potent inhibitor of NF-kappaB activation. Treatment of human myeloid ML-1a cells with tumor necrosis factor rapidly activated NF-kappaB, this activation was completely suppressed by sanguinarine in a dose- and time-dependent manner. Sanguinarine did not inhibit the binding of NF-kappaB protein to the DNA but rather inhibited the pathway leading to NF-kappaB activation. The reversal of inhibitory effects of sanguinarine by reducing agents suggests a critical sulfhydryl group is involved in NF-kappaB activation. Sanguinarine blocked the tumor necrosis factor-induced phosphorylation and degradation of IkappaBalpha, an inhibitory subunit of NF-kappaB, and inhibited translocation of p65 subunit to the nucleus. As sanguinarine also inhibited NF-kappaB activation induced by interleukin-1, phorbol ester, and okadaic acid but not that activated by hydrogen peroxide or ceramide, the pathway leading to NF-kappaB activation is likely different for different inducers. Overall, our results demonstrate that sanguinarine is a potent suppressor of NF-kappaB activation and it acts at a step prior to IkappaBalpha phosphorylation.
Sigma-Aldrich
Sanguinarine chloride hydrate, ≥98% (HPLC)