Merck
  • Elevated Circulating Fetuin-B Levels Are Associated with Insulin Resistance and Reduced by GLP-1RA in Newly Diagnosed PCOS Women.
Elevated Circulating Fetuin-B Levels Are Associated with Insulin Resistance and Reduced by GLP-1RA in Newly Diagnosed PCOS Women.
Mediators of inflammation (2020-10-17)
Mani Mokou, Shan Yang, Bin Zhan, Shan Geng, Kejia Li, Mengliu Yang, Gangyi Yang, Wuquan Deng, Hua Liu, Dongfang Liu, Zhiming Zhu, Ling Li
ABSTRACT
Previous studies have suggested that Fetuin-B seems to be a secreted adipokine related to metabolic diseases. However, the results have been inconsistent. Here, our objective is to investigate the changes in circulating Fetuin-B levels in women with polycystic ovary syndrome (PCOS) and analyze the association of Fetuin-B and insulin resistance (IR). The current study is comprised of a cross-sectional study and a series of interventional studies. Oral glucose tolerance test (OGTT) and euglycemic-hyperinsulinemic clamp (EHC) were engaged to assess glucose tolerance and insulin sensitivity. Serum Fetuin-B levels were determined by ELISA. Serum Fetuin-B and TNF-α levels were markedly increased in women with PCOS compared to healthy women. Circulating Fetuin-B was positively associated with body mass index, waist-to-hip ratio, the percentage of body fat (FAT%), systolic blood pressure, triglyceride, low-density lipoprotein cholesterol, fasting blood glucose, 2 h blood glucose after glucose overload, fasting insulin, 2 h insulin after glucose overload, HOMA-insulin resistance index (HOMA-IR), the area under the curve for insulin (AUCi), AUCg, and TNF-α, while negatively associated with M value and follicular stimulating hormone (FSH). During the EHC, Fetuin-B levels were found to be significantly increased in PCOS women. After a glucose challenge, serum Fetuin-B levels in healthy women were significantly increased. Lipid infusion reduced serum Fetuin-B levels in 30 healthy subjects. After six months of glucagon-like peptide-1 receptor agonist (GLP-1RA) intervention, serum Fetuin-B concentrations in PCOS women markedly decreased following ameliorated IR. Our results indicate that Fetuin-B may be a biomarker of IR in individuals with PCOS. This trial is registered with ChiCTR-IIR-16007901.
Sigma-Aldrich
Human FETUB / Fetuin-B ELISA Kit, for serum, plasma, cell culture supernatants and urine
Pricing and availability is not currently available.