Merck
  • Dimerization of Human Drebrin-like Protein Governs Its Biological Activity.
Dimerization of Human Drebrin-like Protein Governs Its Biological Activity.
Biochemistry (2020-04-14)
Arindam Ghosh, Jörg Enderlein, Eugenia Butkevich
ABSTRACT
Drebrin-like protein (DBNL) is a multidomain F-actin-binding protein, which also interacts with other molecules within different intracellular pathways. Here, we present quantitative measurements on the size and conformation of human DBNL. Using dual-focus fluorescence correlation spectroscopy, we determined the hydrodynamic radius of the DBNL monomer. Native gel electrophoresis and dual-color fluorescence cross-correlation spectroscopy show that both endogenous DBNL and recombinant DBNL exist as dimers under physiological conditions. We demonstrate that C-terminal truncations of DBNL downstream of the coiled-coil domain result in its oligomerization at nanomolar concentrations. In contrast, the ADF-H domain alone is a monomer, which displays a concentration-dependent self-assembly. In vivo FLIM-FRET imaging shows that the presence of only actin-binding domains is not sufficient for DBNL to localize properly at the actin filament inside the cell. In summary, our work provides detailed insight into the structure-function relationship of human drebrin-like protein.
Sigma-Aldrich
Fluoroshield, histology mounting medium
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-DBNL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Cocktail Tablets, EDTA-Free, for use in purification of Histidine-tagged proteins
Pricing and availability is not currently available.