Merck
  • Identification of proteins and cellular pathways targeted by 2-nitroimidazole hypoxic cytotoxins.
Identification of proteins and cellular pathways targeted by 2-nitroimidazole hypoxic cytotoxins.
Redox biology (2021-03-01)
Faisal Bin Rashed, Alexandru Cezar Stoica, Dawn MacDonald, Hassan El-Saidi, Carolynne Ricardo, Bhumi Bhatt, Jack Moore, Diana Diaz-Dussan, Nirilanto Ramamonjisoa, Yvonne Mowery, Sambasivarao Damaraju, Richard Fahlman, Piyush Kumar, Michael Weinfeld
ABSTRACT
Tumour hypoxia negatively impacts therapy outcomes and continues to be a major unsolved clinical problem. Nitroimidazoles are hypoxia selective compounds that become entrapped in hypoxic cells by forming drug-protein adducts. They are widely used as hypoxia diagnostics and have also shown promise as hypoxia-directed therapeutics. However, little is known about the protein targets of nitroimidazoles and the resulting effects of their modification on cancer cells. Here, we report the synthesis and applications of azidoazomycin arabinofuranoside (N3-AZA), a novel click-chemistry compatible 2-nitroimidazole, designed to facilitate (a) the LC-MS/MS-based proteomic analysis of 2-nitroimidazole targeted proteins in FaDu head and neck cancer cells, and (b) rapid and efficient labelling of hypoxic cells and tissues. Bioinformatic analysis revealed that many of the 62 target proteins we identified participate in key canonical pathways including glycolysis and HIF1A signaling that play critical roles in the cellular response to hypoxia. Critical cellular proteins such as the glycolytic enzyme glyceraldehyde-3-phosphate dehydrogenase (GAPDH) and the detoxification enzyme glutathione S-transferase P (GSTP1) appeared as top hits, and N3-AZA adduct formation significantly reduced their enzymatic activities only under hypoxia. Therefore, GAPDH, GSTP1 and other proteins reported here may represent candidate targets to further enhance the potential for nitroimidazole-based cancer therapeutics.
Sigma-Aldrich
Glutathione-S-Transferase (GST) Assay Kit, sufficient for 500 multiwell tests
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Anti-Gstp1 Antibody, serum, from rabbit