Merck
  • Regulation of ALT-associated homology-directed repair by polyADP-ribosylation.
Regulation of ALT-associated homology-directed repair by polyADP-ribosylation.
Nature structural & molecular biology (2020-10-14)
Song My Hoang, Nicole Kaminski, Ragini Bhargava, Jonathan Barroso-González, Michelle L Lynskey, Laura García-Expósito, Justin L Roncaioli, Anne R Wondisford, Callen T Wallace, Simon C Watkins, Dominic I James, Ian D Waddell, Donald Ogilvie, Kate M Smith, Felipe da Veiga Leprevost, Dattatreya Mellacharevu, Alexey I Nesvizhskii, Jianfeng Li, Dominique Ray-Gallet, Robert W Sobol, Genevieve Almouzni, Roderick J O'Sullivan
ABSTRACT
The synthesis of poly(ADP-ribose) (PAR) reconfigures the local chromatin environment and recruits DNA-repair complexes to damaged chromatin. PAR degradation by poly(ADP-ribose) glycohydrolase (PARG) is essential for progression and completion of DNA repair. Here, we show that inhibition of PARG disrupts homology-directed repair (HDR) mechanisms that underpin alternative lengthening of telomeres (ALT). Proteomic analyses uncover a new role for poly(ADP-ribosyl)ation (PARylation) in regulating the chromatin-assembly factor HIRA in ALT cancer cells. We show that HIRA is enriched at telomeres during the G2 phase and is required for histone H3.3 deposition and telomere DNA synthesis. Depletion of HIRA elicits systemic death of ALT cancer cells that is mitigated by re-expression of ATRX, a protein that is frequently inactivated in ALT tumors. We propose that PARylation enables HIRA to fulfill its essential role in the adaptive response to ATRX deficiency that pervades ALT cancers.
Sigma-Aldrich
Anti-Poly ADP-ribose Antibody, clone 10H, clone 10H, from mouse
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, purified from hybridoma cell culture