Merck
  • Dynamic Variations in Brain Glycogen are Involved in Modulating Isoflurane Anesthesia in Mice.
Dynamic Variations in Brain Glycogen are Involved in Modulating Isoflurane Anesthesia in Mice.
Neuroscience bulletin (2020-10-14)
Ze Fan, Zhihao Zhang, Shiyi Zhao, Yuanyuan Zhu, Dong Guo, Bo Yang, Lixia Zhuo, Jiao Han, Rui Wang, Zongping Fang, Hailong Dong, Yan Li, Lize Xiong
ABSTRACT
General anesthesia severely affects the metabolites in the brain. Glycogen, principally stored in astrocytes and providing the short-term delivery of substrates to neurons, has been implicated as an affected molecule. However, whether glycogen plays a pivotal role in modulating anesthesia-arousal remains unclear. Here, we demonstrated that isoflurane-anesthetized mice exhibited dynamic changes in the glycogen levels in various brain regions. Glycogen synthase (GS) and glycogen phosphorylase (GP), key enzymes of glycogen metabolism, showed increased activity after isoflurane exposure. Upon blocking glycogenolysis with 1,4-dideoxy-1,4-imino-D-arabinitol (DAB), a GP antagonist, we found a prolonged time of emergence from anesthesia and an enhanced δ frequency in the EEG (electroencephalogram). In addition, augmented expression of glycogenolysis genes in glycogen phosphorylase, brain (Pygb) knock-in (PygbH11/H11) mice resulted in delayed induction of anesthesia, a shortened emergence time, and a lower ratio of EEG-δ. Our findings revealed a role of brain glycogen in regulating anesthesia-arousal, providing a potential target for modulating anesthesia.
Sigma-Aldrich
1,4-Dideoxy-1,4-imino-D-arabinitol hydrochloride, enzyme inhibitor
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Pricing and availability is not currently available.