Merck
  • Transdermal penetration behaviour of drugs: CART-clustering, QSPR and selection of model compounds.
Transdermal penetration behaviour of drugs: CART-clustering, QSPR and selection of model compounds.
Bioorganic & medicinal chemistry (2007-09-11)
Bram Baert, Eric Deconinck, Mireille Van Gele, Marian Slodicka, Paul Stoppie, Samuel Bodé, Guido Slegers, Yvan Vander Heyden, Jo Lambert, Johan Beetens, Bart De Spiegeleer
ABSTRACT
A set of 116 structurally very diverse compounds, mainly drugs, was characterized by 1630 molecular descriptors. The biological property modelled in this study was the transdermal permeability coefficient logK(p). The main objective was to find a limited set of suitable model compounds for skin penetration studies. The classification and regression trees (CART) approach was applied and the resulting groups were discussed in terms of their role as possible model compounds and their determining descriptors. A second objective was to model transdermal penetration as a function of selected descriptors in quantitative structure-property relationships (QSPR) using a boosted CART (BRT) approach and multiple linear regression (MLR) analysis, where regression models were obtained by stepwise selection of the best descriptors. Evaluation of the standard statistical, as well as descriptor-number dependent, regression quality attributes yielded a maximal 10-dimensional MLR model. The CART and MLR models were subjected to an external validation with a test set of 12 compounds, not included in the original learning set of 104 compounds, to assess the predictive power of the models.
Product Description
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Betamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Thymol, FCC, FG
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Sigma-Aldrich
2-Naphthol, BioXtra, ≥99.0% (GC)
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Naproxen, VETRANAL®, analytical standard
Supelco
Thymol, analytical standard
Sigma-Aldrich
2,4,6-Trichlorophenol, 98%
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Supelco
Ethylene glycol butyl ether, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Thymol, meets analytical specification of Ph. Eur., BP, NF, 99-101%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
2-Naphthol, 99%
Sigma-Aldrich
Anisole, ≥99%, FCC, FG
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
2-Naphthol, 98%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG