Merck
  • TANK-Binding Kinase 1 Regulates the Localization of Acyl-CoA Synthetase ACSL1 to Control Hepatic Fatty Acid Oxidation.
TANK-Binding Kinase 1 Regulates the Localization of Acyl-CoA Synthetase ACSL1 to Control Hepatic Fatty Acid Oxidation.
Cell metabolism (2020-11-06)
Jin Young Huh, Shannon M Reilly, Mohammad Abu-Odeh, Anne N Murphy, Sushil K Mahata, Jinyu Zhang, Yoori Cho, Jong Bae Seo, Chao-Wei Hung, Courtney R Green, Christian M Metallo, Alan R Saltiel, Jin Young Huh, Shannon M Reilly, Mohammad Abu-Odeh, Anne N Murphy, Sushil K Mahata, Jinyu Zhang, Yoori Cho, Jong Bae Seo, Chao-Wei Hung, Courtney R Green, Christian M Metallo, Alan R Saltiel
ABSTRACT
Hepatic TANK (TRAF family member associated NFκB activator)-binding kinase 1 (TBK1) activity is increased during obesity, and administration of a TBK1 inhibitor reduces fatty liver. Surprisingly, liver-specific TBK1 knockout in mice produces fatty liver by reducing fatty acid oxidation. TBK1 functions as a scaffolding protein to localize acyl-CoA synthetase long-chain family member 1 (ACSL1) to mitochondria, which generates acyl-CoAs that are channeled for β-oxidation. TBK1 is induced during fasting and maintained in the unphosphorylated, inactive state, enabling its high affinity binding to ACSL1 in mitochondria. In TBK1-deficient liver, ACSL1 is shifted to the endoplasmic reticulum to promote fatty acid re-esterification in lieu of oxidation in response to fasting, which accelerates hepatic lipid accumulation. The impaired fatty acid oxidation in TBK1-deficient hepatocytes is rescued by the expression of kinase-dead TBK1. Thus, TBK1 operates as a rheostat to direct the fate of fatty acids in hepatocytes, supporting oxidation when inactive during fasting and promoting re-esterification when activated during obesity.
Sigma-Aldrich
MISSION® esiRNA, targeting human ACSL1
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Supelco
Lipid Standard, Mono-, Di-, & Triglyceride Mix, certified reference material, pkg of 40 mg (Neat)
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Palmitoyl-L-carnitine chloride, ≥98% (TLC), powder
Sigma-Aldrich
Coenzyme A sodium salt hydrate, cofactor for acyl transfer
Sigma-Aldrich
Sodium palmitate, ≥98.5%