Merck
  • Single-Cell RNA Sequencing Analysis Reveals Sequential Cell Fate Transition during Human Spermatogenesis.
Single-Cell RNA Sequencing Analysis Reveals Sequential Cell Fate Transition during Human Spermatogenesis.
Cell stem cell (2018-09-04)
Mei Wang, Xixi Liu, Gang Chang, Yidong Chen, Geng An, Liying Yan, Shuai Gao, Yanwen Xu, Yueli Cui, Ji Dong, Yuhan Chen, Xiaoying Fan, Yuqiong Hu, Ke Song, Xiaohui Zhu, Yun Gao, Zhaokai Yao, Shuhui Bian, Yu Hou, Jiahao Lu, Rui Wang, Yong Fan, Ying Lian, Wenhao Tang, Yapeng Wang, Jianqiao Liu, Lianming Zhao, Luyu Wang, Zhaoting Liu, Renpei Yuan, Yujia Shi, Boqiang Hu, Xiulian Ren, Fuchou Tang, Xiao-Yang Zhao, Jie Qiao
ABSTRACT
Spermatogenesis generates mature male gametes and is critical for the proper transmission of genetic information between generations. However, the developmental landscapes of human spermatogenesis remain unknown. Here, we performed single-cell RNA sequencing (scRNA-seq) analysis for 2,854 testicular cells from donors with normal spermatogenesis and 174 testicular cells from one nonobstructive azoospermia (NOA) donor. A hierarchical model was established, which was characterized by the sequential and stepwise development of three spermatogonia subtypes, seven spermatocyte subtypes, and four spermatid subtypes. Further analysis identified several stage-specific marker genes of human germ cells, such as HMGA1, PIWIL4, TEX29, SCML1, and CCDC112. Moreover, we identified altered gene expression patterns in the testicular somatic cells of one NOA patient via scRNA-seq analysis, paving the way for further diagnosis of male infertility. Our work allows for the reconstruction of transcriptional programs inherent to sequential cell fate transition during human spermatogenesis and has implications for deciphering male-related reproductive disorders.
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-CCDC112 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-TJP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SCML1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Stra8 Antibody, from rabbit, purified by affinity chromatography