Merck
  • Phosphorylation of STAT3 by axonal Cdk5 promotes axonal regeneration by modulating mitochondrial activity.
Phosphorylation of STAT3 by axonal Cdk5 promotes axonal regeneration by modulating mitochondrial activity.
Experimental neurology (2020-10-26)
Jinyeon Hwang, Uk Namgung
ABSTRACT
Cyclin-dependent kinase 5 (Cdk5) is involved in neural organization and synaptic functions in developing and adult brains, yet its role in axonal regeneration is not known well. Here, we characterize Cdk5 function for axonal regeneration after peripheral nerve injury. Levels of Cdk5 and p25 were elevated in sciatic nerve axons after injury. Cdk5 activity was concomitantly induced from injured nerve and increased the phosphorylation of signal transducer and activator of transcription 3 (STAT3) on the serine 727 residue. Pharmacological and genetic blockades of Cdk5 activity phosphorylating STAT3 resulted in the inhibition of axonal regeneration as evidenced by reduction of retrograde labeling of dorsal root ganglion (DRG) sensory neurons and spinal motor neurons and also of neurite outgrowth of preconditioned DRG neurons in culture. Cdk5 and STAT3 were found in mitochondrial membranes of the injured sciatic nerve. Cdk5-GFP, which was translocated into the mitochondria by the mitochondrial target sequence (MTS), induced STAT3 phosphorylation in transfected DRG neurons and was sufficient to induce neurite outgrowth. In the mitochondria, Cdk5 activity was positively correlated with increased mitochondrial membrane potential as measured by fluorescence intensity of JC-1 aggregates. Our data suggest that Cdk5 may play a role in modulating mitochondrial activity through STAT3 phosphorylation, thereby promoting axonal regeneration.
Sigma-Aldrich
PP1, ≥98% (HPLC)
Sigma-Aldrich
JC-1, powder or solid (Crystals)
SAFC
Basal Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Digitonin, Used as non-ionic detergent