Merck
  • Temozolomide-Induced RNA Interactome Uncovers Novel LncRNA Regulatory Loops in Glioblastoma.
Temozolomide-Induced RNA Interactome Uncovers Novel LncRNA Regulatory Loops in Glioblastoma.
Cancers (2020-09-16)
Sabrina Fritah, Arnaud Muller, Wei Jiang, Ramkrishna Mitra, Mohamad Sarmini, Monika Dieterle, Anna Golebiewska, Tao Ye, Eric Van Dyck, Christel Herold-Mende, Zhongming Zhao, Francisco Azuaje, Simone P Niclou
ABSTRACT
Resistance to chemotherapy by temozolomide (TMZ) is a major cause of glioblastoma (GBM) recurrence. So far, attempts to characterize factors that contribute to TMZ sensitivity have largely focused on protein-coding genes, and failed to provide effective therapeutic targets. Long noncoding RNAs (lncRNAs) are essential regulators of epigenetic-driven cell diversification, yet, their contribution to the transcriptional response to drugs is less understood. Here, we performed RNA-seq and small RNA-seq to provide a comprehensive map of transcriptome regulation upon TMZ in patient-derived GBM stem-like cells displaying different drug sensitivity. In a search for regulatory mechanisms, we integrated thousands of molecular associations stored in public databases to generate a background "RNA interactome". Our systems-level analysis uncovered a coordinated program of TMZ response reflected by regulatory circuits that involve transcription factors, mRNAs, miRNAs, and lncRNAs. We discovered 22 lncRNAs involved in regulatory loops and/or with functional relevance in drug response and prognostic value in gliomas. Thus, the investigation of TMZ-induced gene networks highlights novel RNA-based predictors of chemosensitivity in GBM. The computational modeling used to identify regulatory circuits underlying drug response and prioritizing gene candidates for functional validation is applicable to other datasets.
Sigma-Aldrich
Mouse IgG1 Negative Control, clone Ci4, PE conjugate, Mouse IgG1 Negative Control Monoclonal Antibody validated for use in Flow Cytometry.
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-phospho-ATM (Ser1981) Antibody, clone 10H11.E12 PE conjugate, clone 10H11.E12, from mouse, PE
Pricing and availability is not currently available.