Merck
  • N-hydroxyurea--a versatile zinc binding function in the design of metalloenzyme inhibitors.
N-hydroxyurea--a versatile zinc binding function in the design of metalloenzyme inhibitors.
Bioorganic & medicinal chemistry letters (2006-06-09)
Claudia Temperini, Alessio Innocenti, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
N-Hydroxyurea binds both to carbonic anhydrase (CA) and to matrix metalloproteinases (MMPs). X-ray crystallography showed N-hydroxyurea to bind in a bidentate mode by means of the oxygen and nitrogen atoms of the NHOH moiety to the Zn(II) ion of CA, participating in a network of hydrogen bonds with a water molecule and Thr199. A derivatized N-hydroxyurea showed low-micromolar affinity for several CAs. This simple zinc binding function may be exploited for obtaining potent metalloenzyme inhibitors, due to its versatility of binding to the metal ion present in the active site of such enzymes.
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Acetohydroxamic acid, 98%