Merck
In silico fragment-based discovery of DPP-IV S1 pocket binders.
Bioorganic & medicinal chemistry letters (2005-12-03)
Christian Rummey, Sonja Nordhoff, Meinolf Thiemann, Günther Metz
ABSTRACT
Dipeptidyl peptidase IV is a clinically validated target for type-2 diabetes and belongs to a family of peptidases with a quite unique post-proline cleavage specificity. Known inhibitors contain a limited number of molecular anchors occupying the small prototypical S1 pocket. A virtual screening approach for such S1-binding fragments was carried out using FlexX docking to evaluate its potential to confirm known and find novel compounds. Several low molecular weight inhibitors exhibiting activities in the micromolar range could be identified as starting points for structure-based design.
Sigma-Aldrich
(1R,2R)-(+)-1,2-Diphenylethylenediamine, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
meso-1,2-Diphenylethylenediamine, 98%
Pricing and availability is not currently available.