Merck
  • Ochratoxin A Levels in Tissues of Wild Boars (Sus scrofa) from Northern Italy.
Ochratoxin A Levels in Tissues of Wild Boars (Sus scrofa) from Northern Italy.
Toxins (2020-11-12)
Tiziano Iemmi, Alessandro Menozzi, Valentina Meucci, Irene Magnini, Federica Battaglia, Lorella Severino, Andrea Ariano, Simone Bertini
ABSTRACT
Ochratoxin A (OTA) is a mycotoxin produced by Aspergillus and Penicillium, capable of contaminating several foodstuffs. OTA damages primarily the kidneys, and is suspected to be a carcinogenic substance, thus maximum levels for OTA in foodstuffs have been established in the EU. Italian Ministry of Health suggested a maximum level of 1 μg/kg OTA in pork meat and derived products. In this study, OTA concentrations in liver, kidney, and muscle of 64 wild boars (Sus scrofa) killed in two areas (area A and B) of Parma province (northern Italy), characterized by different habitat types, were assessed by HPLC-FLD technique. OTA was detected in 54% liver, 52% kidney, and 16% muscle samples. OTA levels were significantly higher in liver and kidney compared with muscle, and were above 1 μg/kg in 19 liver, 17 kidney, and 4 muscle samples. OTA levels in wild boars from area A resulted significantly higher with respect to those from area B, suggesting an environmental influence on OTA contamination in wild boars. This study seems to confirm that wild boar meat is a potential source of OTA, thus monitoring the presence of this mycotoxin in game meat might be recommended to prevent risks for human health.
Product Description
Supelco
Ochratoxin B solution, ~10 μg/mL in acetonitrile, analytical standard
Pricing and availability is not currently available.