Merck
  • Activity-Dependent Secretion of Synaptic Organizer Cbln1 from Lysosomes in Granule Cell Axons.
Activity-Dependent Secretion of Synaptic Organizer Cbln1 from Lysosomes in Granule Cell Axons.
Neuron (2019-05-11)
Keiji Ibata, Maya Kono, Sakae Narumi, Junko Motohashi, Wataru Kakegawa, Kazuhisa Kohda, Michisuke Yuzaki
ABSTRACT
Synapse formation is achieved by various synaptic organizers. Although this process is highly regulated by neuronal activity, the underlying molecular mechanisms remain largely unclear. Here we show that Cbln1, a synaptic organizer of the C1q family, is released from lysosomes in axons but not dendrites of cerebellar granule cells in an activity- and Ca2+-dependent manner. Exocytosed Cbln1 was retained on axonal surfaces by binding to its presynaptic receptor neurexin. Cbln1 further diffused laterally along the axonal surface and accumulated at boutons by binding postsynaptic δ2 glutamate receptors. Cbln1 exocytosis was insensitive to tetanus neurotoxin, accompanied by cathepsin B release, and decreased by disrupting lysosomes. Furthermore, overexpression of lysosomal sialidase Neu1 not only inhibited Cbln1 and cathepsin B exocytosis in vitro but also reduced axonal bouton formation in vivo. Our findings imply that co-release of Cbln1 and cathepsin B from lysosomes serves as a new mechanism of activity-dependent coordinated synapse modification.
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Earle′s Balanced Salts, With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Lysosome Isolation Kit, sufficient for 25 g (tissue), sufficient for 20 mL (packed cells), enrichment of lysosomes from tissues and packed cells
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, HRP-conjugate, 1 mg/mL, Upstate®
Roche
DNase I, from bovine pancreas
Sigma-Aldrich
Putrescine dihydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Insulin from bovine pancreas, γ-irradiated, BioXtra, suitable for cell culture, potency: ≥20 units/mg (USP units), lyophilized powder
Sigma-Aldrich
apo-Transferrin human, powder, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid
Sigma-Aldrich
Anti-MAP2 (2a+2b) antibody, Mouse monoclonal, clone AP-20, ascites fluid
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, HRP conjugate, Upstate®, from goat
Sigma-Aldrich
Xylazine hydrochloride, ≥99.0% (HPLC)
Sigma-Aldrich
Tetanus toxin from Clostridium tetani