Merck
  • A label-free impedance assay in endothelial cells differentiates the activation and desensitization properties of clinical S1P1 agonists.
A label-free impedance assay in endothelial cells differentiates the activation and desensitization properties of clinical S1P1 agonists.
FEBS open bio (2020-08-19)
Patrick Grailhe, Asma Boutarfa-Madec, Philippe Beauverger, Philip Janiak, Ashfaq A Parkar
ABSTRACT
Sphingosine-1 phosphate receptor-1 (S1P1 ) activation maintains endothelial barrier integrity, whereas S1P1 desensitization induces peripheral blood lymphopenia. The latter is exploited in the approval and/or late-stage development of receptor-desensitizing agents targeting the S1P1 receptor in multiple sclerosis, such as siponimod, ozanimod, and ponesimod. SAR247799 is a recently described G protein-biased S1P1 agonist that activates S1P1 without desensitization and thus has endothelial-protective properties in patients without reducing lymphocytes. As SAR247799 demonstrated endothelial-protective effects at sub-lymphocyte-reducing doses, the possibility exists that other S1P1 modulators could also exhibit endothelial-protective properties at lower doses. To explore this possibility, we sought to quantitatively compare the biased properties of SAR247799 with the most advanced clinical molecules targeting S1P1 . In this study, we define the β-arrestin pathway component of the impedance profile following S1P1 activation in a human umbilical vein endothelial cell line (HUVEC) and report quantitative indices of the S1P1 activation-to-desensitization ratio of various clinical molecules. In a label-free impedance assay assessing endothelial barrier integrity and disruption, the mean estimates (95% confidence interval) of the activation-to-desensitization ratios of SAR247799, ponesimod, ozanimod, and siponimod were 114 (91.1-143), 7.66 (3.41-17.2), 6.35 (3.21-12.5), and 0.170 (0.0523-0.555), respectively. Thus, we show that SAR247799 is the most G protein-biased S1P1 agonist currently characterized. This rank order of bias among the most clinically advanced S1P1 modulators provides a new perspective on the relative potential of these clinical molecules for improving endothelial function in patients in relation to their lymphocyte-reducing (desensitization) properties.
Sigma-Aldrich
1H-Indazole-5-carboxylic acid, 97%
Pricing and availability is not currently available.