Merck
  • Macrophage secretion of miR-106b-5p causes renin-dependent hypertension.
Macrophage secretion of miR-106b-5p causes renin-dependent hypertension.
Nature communications (2020-09-25)
J Oh, S J Matkovich, A E Riek, S M Bindom, J S Shao, R D Head, R A Barve, M S Sands, G Carmeliet, P Osei-Owusu, R H Knutsen, H Zhang, K J Blumer, C G Nichols, R P Mecham, Á Baldán, B A Benitez, M L Sequeira-Lopez, R A Gomez, C Bernal-Mizrachi
ABSTRACT
Myeloid cells are known mediators of hypertension, but their role in initiating renin-induced hypertension has not been studied. Vitamin D deficiency causes pro-inflammatory macrophage infiltration in metabolic tissues and is linked to renin-mediated hypertension. We tested the hypothesis that impaired vitamin D signaling in macrophages causes hypertension using conditional knockout of the myeloid vitamin D receptor in mice (KODMAC). These mice develop renin-dependent hypertension due to macrophage infiltration of the vasculature and direct activation of renal juxtaglomerular (JG) cell renin production. Induction of endoplasmic reticulum stress in knockout macrophages increases miR-106b-5p secretion, which stimulates JG cell renin production via repression of transcription factors E2f1 and Pde3b. Moreover, in wild-type recipient mice of KODMAC/miR106b-/- bone marrow, knockout of miR-106b-5p prevents the hypertension and JG cell renin production induced by KODMAC macrophages, suggesting myeloid-specific, miR-106b-5p-dependent effects. These findings confirm macrophage miR-106b-5p secretion from impaired vitamin D receptor signaling causes inflammation-induced hypertension.
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Pricing and availability is not currently available.
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Mouse Ren1 / Renin-1 ELISA Kit, for serum, plasma and cell culture supernatants
Pricing and availability is not currently available.
Sigma-Aldrich
Rat Renin-1 ELISA Kit, for cell culture supernatants, plasma, and serum samples
Pricing and availability is not currently available.