Merck
  • Splicing factor PRPF6 upregulates oncogenic androgen receptor signaling pathway in hepatocellular carcinoma.
Splicing factor PRPF6 upregulates oncogenic androgen receptor signaling pathway in hepatocellular carcinoma.
Cancer science (2020-08-04)
Huijuan Song, Ning Sun, Lin Lin, Shan Wei, Kai Zeng, Wei Liu, Chunyu Wang, Xinping Zhong, Manlin Wang, Shengli Wang, Baosheng Zhou, Chi Lv, Wensu Liu, Yue Zhao
ABSTRACT
Androgen receptor (AR) signaling is considered to be crucial for the pathogenesis of hepatocellular carcinoma (HCC) with obvious sexual dimorphism. Pre-mRNA processing factor 6 (PRPF6) was identified as a coactivator of AR. However, the molecular mechanism underlying the modulation function of PRPF6 on AR-mediated transcriptional activity in HCC needs to be further clarified. In this study, we analyzed data from The Cancer Genome Atlas to show that PRPF6 is highly expressed in HCC. . Our data indicated that PRPF6 interacts with AR/AR splice variants (AR-Vs) and upregulates AR/AR splice variant 7-mediated transcriptional activity even without dihydrotestosterone treatment. We observed that AR is obviously induced by androgen treatment and is mainly expressed in the nucleus in HCC-derived cell lines. Moreover, overexpression of PRPF6 enhances AR expression accompanied with the increase of AR-Vs expression. We provided evidence that PRPF6 participates in upregulating AR self-transcription. PRPF6 facilitates the recruitment of AR to the androgen responsive element region of the AR gene. Finally, PRPF6 depletion inhibits cell proliferation in HCC cells and mouse xenografts. Taken together, our results suggest that PRPF6 as a splicing factor enhances AR self-transcription, thereby coactivating oncogenic AR/AR-Vs actions in HCC.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys36) Antibody, from rabbit, purified by affinity chromatography
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-CDK20 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab3
Pricing and availability is not currently available.