Merck
  • Effect of Drying Methods on Bioactive Compounds and Antioxidant Capacity in Grape Skin Residues from the New Hybrid Variety "BRS Magna".
Effect of Drying Methods on Bioactive Compounds and Antioxidant Capacity in Grape Skin Residues from the New Hybrid Variety "BRS Magna".
Molecules (Basel, Switzerland) (2020-08-23)
Gabriela Viana da Silva, Bruna Aparecida Souza Machado, Walkia Polliana de Oliveira, Camilla Fernanda Godinho da Silva, Cedenir Pereira de Quadros, Janice Izabel Druzian, Ederlan de Souza Ferreira, Marcelo Andrés Umsza-Guez
ABSTRACT
The effects of the drying process using the conventional oven and freeze-drying on the thermogravimetric profile, proximate composition, color parameters, individual bioactive compounds, and antioxidant activity in the grape residue (skin) were evaluated. Twenty individual phenolic compounds were identified, where a variation in concentration was observed for flavonols, stilbenes, phenolic acids, flavonoids, procyanidins, and particularly anthocyanins (malvidin-3,5-di-O-glucoside of 253.2-536.9 mg/kg) due to the drying process. Drying in a conventional oven caused a decrease of 23% of the total polyphenols. The skin of the BRS magna grape has a high concentration of total phenolic content of 489.5-148.3 mg.GAE/100 g, total anthocyanin content of 124.9-260.1 mg.CE/100 g, and total flavonoid content of 12.7-26.0 mg.QE/100 g. The results of free radical scavenging activity (1.26-4.91 μg/mL, as EC50) and ferric reducing antioxidant power (82.93-184.10 μmol/g of skin as equivalent to Fe2SO4) indicate high antioxidant activity, independently of the drying process applied. It was concluded that, if the application is directed to anthocyanin compounds, the use of lyophilization is recommended. On the other hand, if the interest is in bioactive compounds that exert antioxidant activity, conventional oven-drying can be used.
Sigma-Aldrich
Cyanin chloride, ≥90% (HPLC)
Pricing and availability is not currently available.
Rutin, phyproof® Reference Substance
Pricing and availability is not currently available.