Merck
  • Dielectric Relaxation Characteristics of Epoxy Resin Modified with Hydroxyl-Terminated Nitrile Rubber.
Dielectric Relaxation Characteristics of Epoxy Resin Modified with Hydroxyl-Terminated Nitrile Rubber.
Molecules (Basel, Switzerland) (2020-09-16)
Chi Chen, Qing Sun, Chuang Wang, Yue Bu, Jiawei Zhang, Zongren Peng
ABSTRACT
Utilizing liquid rubber to toughen epoxy resin is one of the most mature and promising methods. However, the dielectric relaxation characteristics of the epoxy/liquid rubber composites have not been studied systematically, while the relaxation behaviours are a critical factor for both micro and macro properties. In this paper, hydroxyl-terminated liquid nitrile rubber (HTBN) is employed to reinforce a kind of room-temperature-cured epoxy resin. The dielectric spectrum is measured and analysed. Results show that two relaxation processes are introduced in the binary composites. The α relaxation of HTBN shows a similar temperature dependence with the β relaxation of epoxy resin. The interfacial polarization leads to an increase of complex permittivity, which reaches its maximum at 70 °C. In addition, affected by interfacial polarization, the thermionic polarization is inhibited, and the samples with filler ratios of 15% and 25% show lower DC-conductivity below 150 °C. In addition, the α relaxation and thermionic polarization of epoxy resin obey the Vogel‒Fulcher‒Tammann law, while the interfacial polarization and DC-conductivity satisfy with the Arrhenius law. Furthermore, the fitting results of the Vogel temperature of α relaxation, glass transition temperature, apparent activation energy of interfacial polarization and DC-conductivity all decline with HTBN content. These results can provide a reference and theoretical guidance for the assessment of dielectric properties and the improvement of the formulation of liquid-rubber-toughened epoxy resin.
Sigma-Aldrich
1,2-Diaminopropane, 99%
Pricing and availability is not currently available.