Merck
  • Utilising Magnetically Isolated Lysosomes for Direct Quantification of Intralysosomal Drug Concentrations by LC-MS/MS Analysis: An Investigatory Study With Imipramine.
Utilising Magnetically Isolated Lysosomes for Direct Quantification of Intralysosomal Drug Concentrations by LC-MS/MS Analysis: An Investigatory Study With Imipramine.
Journal of pharmaceutical sciences (2020-06-07)
Laura Francis, Andrew Harrell, David Hallifax, Aleksandra Galetin
ABSTRACT
Lysosomes are acidic intracellular organelles that can extensively sequester basic lipophilic drugs due to pH and membrane partitioning, and therefore may significantly influence subcellular drug concentrations. Current in vitro methods for lysosomal drug sequestration evaluation typically lack the ability to accurately and sensitively quantify drug concentrations directly within the lysosome. In the current study, magnetic lysosomal isolation was used in the lysosome rich rat NR8383 cell line and combined with LC-MS/MS analysis to quantify intralysosomal concentrations and lysosomal partitioning (KpLysosome) values of imipramine. The purity of the isolated lysosomes was validated by enzymatic and electron microscopy analysis. Lysosomal imipramine accumulation was explored using 2 methods: addition of imipramine to cells followed by lysosomal isolation (Method 1), and direct addition of imipramine to isolated lysosomes (Method 2). This work highlighted that both experimental buffers and ATP influence intralysosomal drug concentrations, and non-specific drug binding and re-distribution limits the use of Method 1. Method 2 may benefit future lysosomal drug accumulation studies, as imipramine demonstrated high KpLysosome values (3500), comparable to in silico predictions. This study reports a novel method for the direct quantification of intralysosomal drug concentrations that has the ability to be adapted to other cell types.
Sigma-Aldrich
β-N-Acetylglucosaminidase Assay Kit, sufficient for 50 reactions (1 mL), sufficient for 500 reactions (100 μL)
Sigma-Aldrich
Magnesium acetate solution, BioUltra, for molecular biology, ~1 M in H2O