Merck
  • The identification, characterization, and distribution of guinea pig CCR4 and epitope mapping of a blocking antibody.
The identification, characterization, and distribution of guinea pig CCR4 and epitope mapping of a blocking antibody.
The Journal of biological chemistry (2001-11-29)
Louise A Jopling, Ian Sabroe, David P Andrew, Tracey J Mitchell, You Li, Martin R Hodge, Timothy J Williams, James E Pease
ABSTRACT
Th2 lymphocytes play a central role in the control and maintenance of allergic inflammation. The chemokine receptor CCR4 is preferentially expressed on the surface of Th2 lymphocytes polarised in vitro. However, CCR4 is found on the surface of a significant proportion of circulating memory T lymphocytes, some of which are capable of producing the Th1-associated cytokine interferon gamma. To investigate the function of CCR4 on guinea pig (gp) T lymphocytes, we identified the open-reading frame of gpCCR4, which encodes a 361-amino acid protein with 88 and 81% amino acid identity to human and murine CCR4 sequences, respectively. Cells transfected with gpCCR4 migrated toward the human and murine orthologues of the CCR4 ligands, macrophage-derived chemokine and thymus and activation-regulated chemokine. Surface expression of CCR4, using an anti-human CCR4 monoclonal antibody, 10E4, was detected on approximately 12% of guinea pig peripheral blood T helper cells, and CCR4(+) guinea pig thymocytes were detected in low numbers. However, CCR4(+) T helper cells constituted approximately 9% of the T lymphocyte population within the normal guinea pig lung and 52% of the guinea pig bronchoalveolar lavage fluid, which is consistent with a role for CCR4 in T lymphocyte development and trafficking through normal tissues. Subsequent analysis of chimeric chemokine receptors indicated that 10E4, a functional inhibitor of gpCCR4 responses, recognized the amino terminus of CCR4.
Sigma-Aldrich
Mouse IgG2a Isotype Control from murine myeloma, clone UPC-10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Mouse IgG1-k Negative Control, clone MOPC-21, Azide Free Antibody, clone MOPC-21, from mouse