Merck
  • ATR Restrains DNA Synthesis and Mitotic Catastrophe in Response to CDC7 Inhibition.
ATR Restrains DNA Synthesis and Mitotic Catastrophe in Response to CDC7 Inhibition.
Cell reports (2020-09-03)
Michael D Rainey, Declan Bennett, Rachel O'Dea, Melania E Zanchetta, Muriel Voisin, Cathal Seoighe, Corrado Santocanale
ABSTRACT
DNA replication initiates from multiple origins, and selective CDC7 kinase inhibitors (CDC7is) restrain cell proliferation by limiting origin firing. We have performed a CRISPR-Cas9 genome-wide screen to identify genes that, when lost, promote the proliferation of cells treated with sub-efficacious doses of a CDC7i. We have found that the loss of function of ETAA1, an ATR activator, and RIF1 reduce the sensitivity to CDC7is by allowing DNA synthesis to occur more efficiently, notably during late S phase. We show that partial CDC7 inhibition induces ATR mainly through ETAA1, and that if ATR is subsequently inhibited, origin firing is unleashed in a CDK- and CDC7-dependent manner. Cells are then driven into a premature and highly defective mitosis, a phenotype that can be recapitulated by ETAA1 and TOPBP1 co-depletion. This work defines how ATR mediates the effects of CDC7 inhibition, establishing the framework to understand how the origin firing checkpoint functions.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
5-Iodo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
RO-3306, ≥98% (HPLC)
Sigma-Aldrich
Roscovitine, ≥98% (TLC)
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, powder, suitable for cell culture
Sigma-Aldrich
N-Lauroylsarcosine sodium salt solution, 20%, for molecular biology
Sigma-Aldrich
Fast Green FCF, Dye content ≥85 %
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
5-Chloro-2′-deoxyuridine, thymidine analog
Sigma-Aldrich
Anti-DNA Antibody, single stranded, clone 16-19, clone 16-19, Chemicon®, from mouse
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
PP1, ≥98% (HPLC)