Merck
  • Human cells adapt to translational errors by modulating protein synthesis rate and protein turnover.
Human cells adapt to translational errors by modulating protein synthesis rate and protein turnover.
RNA biology (2019-10-02)
Ana Sofia Varanda, Mafalda Santos, Ana R Soares, Rui Vitorino, Patrícia Oliveira, Carla Oliveira, Manuel A S Santos
ABSTRACT
Deregulation of tRNAs, aminoacyl-tRNA synthetases (aaRS) or tRNA modifying enzymes, increase the level of protein synthesis errors (PSE) and are associated with several diseases, but the cause-effect mechanisms of these pathologies remain elusive. To clarify the role of PSE in human biology, we have engineered a HEK293 cell line to overexpress a wild type (Wt) tRNASer and two tRNASer mutants that misincorporate serine at non-cognate codon sites. Then, we followed long-term adaptation to PSE of such recombinant cells by analysing cell viability, protein synthesis rate and activation of protein quality control mechanisms (PQC). Engineered cells showed higher level of misfolded and aggregated proteins; activated the ubiquitin-proteasome system (UPS) and the unfolded protein response (UPR), indicative of proteotoxic stress. Adaptation to PSE involved increased protein turnover, UPR up-regulation and altered protein synthesis rate. Gene expression analysis showed that engineered cells presented recurrent alterations in the endoplasmic reticulum, cell adhesion and calcium homeostasis. Herein, we unveil new phenotypic consequences of protein synthesis errors in human cells and identify the protein quality control processes that are necessary for long-term adaptation to PSE and proteotoxic stress. Our data provide important insight on how chronic proteotoxic stress may cause disease and highlight potential biological pathways that support the association of PSE with disease.
Sigma-Aldrich
N-Succinyl-Leu-Leu-Val-Tyr-7-amido-4-trifluoromethylcoumarin, ≥98% (HPLC), solid
Pricing and availability is not currently available.
Sigma-Aldrich
Triton X-100, laboratory grade
Pricing and availability is not currently available.
Sigma-Aldrich
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Brilliant Blue G, pure
Pricing and availability is not currently available.
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
MG-132(R), ≥95% (HPLC)
Pricing and availability is not currently available.
Roche
Cell Proliferation ELISA, BrdU (colorimetric), sufficient for ≤1,000 tests
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-Ubiquitin antibody produced in mouse, clone 6C1, ascites fluid
Pricing and availability is not currently available.