Merck
  • Acute aquatic toxicity of sulfur mustard and its degradation products to Daphnia magna.
Acute aquatic toxicity of sulfur mustard and its degradation products to Daphnia magna.
Marine environmental research (2020-08-28)
Michał Czub, Jakub Nawała, Stanisław Popiel, Daniel Dziedzic, Tomasz Brzeziński, Piotr Maszczyk, Hans Sanderson, Jacek Fabisiak, Jacek Bełdowski, Lech Kotwicki
ABSTRACT
Sulphur mustard (HD) was the most widely produced chemical warfare agent (CWA) in the history of chemical warfare (CW). Simultaneously, the loads of HD account as by far the largest fraction of the sea-dumped CW. Nowadays its presence in the marine ecosystems recognized as a serious threat for marine users and maritime industries. Although, during over a decade of research much has been done to assess the environmental threats linked with underwater chemical munitions. There are, however, essential gaps in scientific knowledge including scarce information about the aquatic toxicity thresholds of HD and its degradation products. Standardized biotests were performed according to the Organisation for Economic Co-operation and Development (OECD) Test No. 202: Daphnia sp. Acute Immobilisation Test guidelines. Obtained results provide a solid foundation for comparison and categorisation of threats of HD and its degradation products. With the D. magna LC50 aquatic acute toxicity threshold at as low as 224 ± 12 μg × L-1, 1,2,5-trithiepane is very toxic, being one of the most toxic CWA degradation products that have been investigated up to date. It exhibits stronger effects than 1,4,5-oxadithiepane and diluted HD that turn out to be toxic. In total, the toxicity of 7 compounds has been estimated. Whenever possible, toxicity thresholds were compared with previously existing data originating from different biotests and mathematical modelling.
Sigma-Aldrich
1,4-Thioxane, 98%
Pricing and availability is not currently available.