Merck
  • Inhibition of T cell activation and function by the adaptor protein CIN85.
Inhibition of T cell activation and function by the adaptor protein CIN85.
Science signaling (2019-02-07)
Mei Suen Kong, Akiko Hashimoto-Tane, Yusuke Kawashima, Machie Sakuma, Tadashi Yokosuka, Kohei Kometani, Reiko Onishi, Nick Carpino, Osamu Ohara, Tomohiro Kurosaki, Kia Kien Phua, Takashi Saito
ABSTRACT
T cell activation is initiated by signaling molecules downstream of the T cell receptor (TCR) that are organized by adaptor proteins. CIN85 (Cbl-interacting protein of 85 kDa) is one such adaptor protein. Here, we showed that CIN85 limited T cell responses to TCR stimulation. Compared to activated wild-type (WT) T cells, those that lacked CIN85 produced more IL-2 and exhibited greater proliferation. After stimulation of WT T cells with their cognate antigen, CIN85 was recruited to the TCR signaling complex. Early TCR signaling events, such as phosphorylation of ζ-chain-associated protein kinase 70 (Zap70), Src homology 2 (SH2) domain-containing leukocyte protein of 76 kDa (SLP76), and extracellular signal-regulated kinase (Erk), were enhanced in CIN85-deficient T cells. The inhibitory function of CIN85 required the SH3 and PR regions of the adaptor, which associated with the phosphatase suppressor of TCR signaling-2 (Sts-2) after TCR stimulation. Together, our data suggest that CIN85 is recruited to the TCR signaling complex and mediates inhibition of T cell activation through its association with Sts-2.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Monoclonal ANTI-FLAG® M1 antibody, clone M1, purified immunoglobulin, buffered aqueous solution
Millipore
HIS-Select® Nickel Magnetic Agarose Beads