Merck
  • A multiplex TaqMan qPCR assay for sensitive and rapid detection of phytoplasmas infecting Rubus species.
A multiplex TaqMan qPCR assay for sensitive and rapid detection of phytoplasmas infecting Rubus species.
PloS one (2017-05-26)
Holger Linck, Erika Krüger, Annette Reineke
ABSTRACT
Rubus stunt is an economically important disease in the production of raspberries, blackberries, and loganberries. A fast, sensitive, and reliable diagnosis of phytoplasmas, the causal agent of the disease, is of prime importance to stop its spread by vegetative propagation and by insect vectors. Therefore, multiplex qPCR assays using TaqMan probes with different kinds of fluorophores in one reaction were developed, allowing the detection of phytoplasmas in general as well as a more specific detection of phytoplasmas belonging to group 16SrV and host DNA (either plant or insect). This assay now provides a practical tool for the screening of motherplants and monitoring the presence and distribution of phytoplasmas in Rubus plants of different geographic origins, cultivars, and cultivation systems, as well as in putative insect vectors like leafhoppers.
Product Description
Roche
KAPA PROBE FAST, suitable for qPCR, 2 ×, Universal
Pricing and availability is not currently available.