Merck
Rescue of Pink1 Deficiency by Stress-Dependent Activation of Autophagy.
Cell chemical biology (2017-04-04)
Yuxi Zhang, David T Nguyen, Ellen M Olzomer, Gin P Poon, Nicholas J Cole, Anita Puvanendran, Brigitte R Phillips, Daniel Hesselson
ABSTRACT
Stimulating autophagy is a promising therapeutic strategy for slowing the progression of neurodegenerative disease. Neurons are insensitive to current approaches based on mTOR inhibition for activating autophagy, and instead may rely on the Parkinson's disease-associated proteins PINK1 and PARKIN to activate the autophagy-lysosomal pathway in response to mitochondrial damage. We developed a multifactorial zebrafish drug-screening platform combining Pink1 deficiency with an environmental toxin to compromise mitochondrial function and trigger dopaminergic neuron loss. Using a phenotypic screening strategy, we identified a series of piperazine phenothiazines, including trifluoperazine, which rescued Pink1 deficiency by activating autophagy selectively in stressed zebrafish and human cells. We show that trifluoperazine acts downstream of, or parallel to, PINK1/PARKIN to stimulate transcription factor EB nuclear translocation and the expression of autophagy-lysosomal target genes. These data suggest that stress-dependent pharmacological reactivation of autophagy could prevent the loss of vulnerable neurons to slow neurodegeneration.
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Mitochondria Staining Kit, 1 kit sufficient for 40 tests (of 5 mL cell suspensions), 1 kit sufficient for 200 tests (of 1 mL cell suspensions)
Roche
KAPA SYBR® FAST One-Step, suitable for RT-PCR, 2 ×, Universal
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder