Merck
  • Increased LGALS3BP promotes proliferation and migration of oral squamous cell carcinoma via PI3K/AKT pathway.
Increased LGALS3BP promotes proliferation and migration of oral squamous cell carcinoma via PI3K/AKT pathway.
Cellular signalling (2019-07-16)
Xiaoxin Zhang, Haoyue Ding, Zhanyi Lu, Liang Ding, Yuxian Song, Yue Jing, Qingang Hu, Yingchun Dong, Yanhong Ni
ABSTRACT
Previous studies showed that lectin galactoside-binding soluble 3 binding protein (LGALS3BP) is an important participant in tumor progression. However, its prognostic value and functional mechanism in oral squamous cell carcinoma (OSCC) are still unclear. In this study, we analyzed LGALS3BP expression in OSCC tissues via Oncomine databases and immunohistochemical staining. LGALS3BP was significantly up-regulated in OSCC tumor tissues. IHC analysis showed that LGALS3BP was predominantly expressed in tumor cells and correlated with poor clinical characteristics. In addition, high LGALS3BP expression predicted poor clinical outcomes and multivariate analysis revealed that LGALS3BP expression was as an independent prognostic factor for OS, DFS and RFS (p < .0001, p = .002, p = .002). Mechanically, LGALS3BP regulated OSCC proliferation and migration via PI3K/AKT pathways, which was abrogated by PI3K inhibitor LY294002 in a dose-dependent manner. Our results suggested that LGALS3BP could be served as a novel independent prognostic factor as well as a potential therapeutic target for OSCC treatment.