Merck
  • Genetic manipulation of primary human natural killer cells to investigate the functional and oncogenic roles of PRDM1.
Genetic manipulation of primary human natural killer cells to investigate the functional and oncogenic roles of PRDM1.
Haematologica (2020-08-01)
Gehong Dong, Yuping Li, Logan Lee, Xuxiang Liu, Yunfei Shi, Xiaoqian Liu, Alyssa Bouska, Qiang Gong, Lingbo Kong, Jinhui Wang, Chih-Hong Lou, Timothy W McKeithan, Javeed Iqbal, Wing C Chan
ABSTRACT
Extra-nodal natural killer/T-cell lymphoma, nasal type (ENKTCL) is a highly aggressive lymphoma, where the tumor suppressor gene (TSG) PRDM1 is frequently lost/inactivated. We employed two different CRISPR/Cas9 approaches to generate PRDM1-/- primary NK cells to study its role in NK-cell homeostasis. PRDM1-/- NK cells showed a marked increase in cloning efficiency, higher proliferation rate and less apoptosis compared with their wild type counterparts. Gene expression profiling demonstrated a marked enrichment in pathways associated with proliferation, cell cycle, MYC, MYB and TCR/NK signaling in PRDM1-/- NK cells, but pathways associated with normal cellular functions including cytotoxic functions were down-regulated, suggesting that the loss of PRDM1 shifted NK cells toward proliferation and survival rather than the performance of its normal functions. We were also able to further modify a PRDM1 deleted clone to introduce heterozygous deletions of common TSG in ENKTCL such as TP53, DDX3X, or PTPN6. We have established an in vitro model to elucidate the major pathways through which PRDM1 mediates its homeostatic control of NK-cells. This approach can be applied to the study of other relevant genetic lesions and oncogenic collaborations in lymphoma pathogenesis.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution