- Structure and mechanism of the RNA polymerase II transcription machinery.
Structure and mechanism of the RNA polymerase II transcription machinery.
Advances in structure determination of the bacterial and eukaryotic transcription machinery have led to a marked increase in the understanding of the mechanism of transcription. Models for the specific assembly of the RNA polymerase II transcription machinery at a promoter, conformational changes that occur during initiation of transcription, and the mechanism of initiation are discussed in light of recent developments.