Merck
  • Influence of salinity stress on bacterial community composition and β-glucosidase activity in a tropical estuary: Elucidation through microcosm experiments.
Influence of salinity stress on bacterial community composition and β-glucosidase activity in a tropical estuary: Elucidation through microcosm experiments.
Marine environmental research (2020-07-15)
Ranjith Eswaran, Lidita Khandeparker
ABSTRACT
The influence of changing salinity on community composition and functional activity (Bacterial Production (BP) and ectoenzyme activity) of major bacterial taxa was evaluated using microcosm experiments in a tropical monsoon influenced estuary. Natural bacterial inocula at different salinities, representing marine, brackish, and freshwater, were inter-transferred and elucidated their response with an emphasis on community composition and β-Glucosidase (BGase) activity. The results revealed a significant decrease in the total bacterial count (TBC) and BP on the translocation of bacterial inocula to different salinity conditions in the case of freshwater bacteria. However, a significant increase in BGase activity coupled with shifts in the studied bacterial groups was evident in the case of marine as well as freshwater bacteria. Quantitative PCR (qPCR) revealed a shift in major bacterial taxa upon translocation to different waters, which was dependent on salinity and the source of inocula. Redundancy and qPCR analyses showed that members belonging to Gammaproteobacteria and Betaproteobacteria were higher, and possibly influenced BGase activity in marine and freshwater, respectively. Translocation of marine inocula to brackish and freshwater resulted in an emergence of Bacteroidetes, Actinobacteria, and Betaproteobacteria, respectively. Whereas, when freshwater inocula were translocated to marine or brackish water, Alphaproteobacteria and Gammaproteobacteria taxa emerged, and this was coupled with increased BGase activity. In contrast, brackish water bacteria showed a strong persistence in bacterial community composition when translocated to different salinities within this estuary. Such phylogenetic persistence or changes suggests species level shifts in specific bacterial taxa, and unravelling the same using different functional gene markers would ascertain their role in organic matter processing and is way forward.
Product Description
Millipore
MCE Membrane Filter, 0.22 μm Pore Size, MF-Millipore, filter diam. 47 mm, hydrophilic, white