Merck
  • Effect of Ginsenoside-Rh2 and Curcurbitacin-B on Cryptosporidium parvum in vitro.
Effect of Ginsenoside-Rh2 and Curcurbitacin-B on Cryptosporidium parvum in vitro.
Experimental parasitology (2020-03-14)
Md Shahiduzzaman, Refaat Ras, Giovanni Widmer
ABSTRACT
Ginsenoside-Rh2 and cucurbitacin-B (CuB) are secondary metabolites of Ginseng (Panax ginseng) and Cucurbitaceae plants respectively. We assessed the anticryptosporidial activity of these two functional compounds in a cell culture model of cryptosporidiosis. The highest concentration of each compound that was not toxic to the host cells was used to assess the activity against C. parvum during infection/invasion and growth in HCT-8 cell monolayers. Monolayers were infected with pre-excysted C. parvum oocysts. Infected monolayers were incubated at 37 °C for 24 h and 48 h in the presence of different concentrations of each test compound. A growth resumption assay was performed by incubating infected monolayers in the presence of compounds for 24 h followed by a second 24-h incubation in the absence of compound. To screen for invasion inhibiting activity, freshly excysted C. parvum sporozoites were pre-treated with different concentrations of compounds prior to adding them to the cell monolayers. Paromomycin, a known inhibitor of C. parvum, and DMSO were used as positive and negative control, respectively. The level of infection was initially assessed using an immunofluorescent assay and quantified by real-time PCR. Both compounds were found to strongly inhibit C. parvum intracellular development in a dose-dependent manner. IC50 values of 25 μM for a 24 h development period and 5.52 μM after 48 h development were measured for Rh2, whereas for CuB an IC50 value of 0.169 μg/ml and 0.118 μg/ml were obtained for the same incubation periods. CuB also effectively inhibited resumption of growth, an activity that was not observed with Rh2. CuB was more effective at inhibiting excystation and/or host cell invasion, indicating that this compound also targets extracellular stages of the parasite.