Merck
  • pH-Dependent Chiral Recognition of D- and L-Arginine Derived Polyamidoamino Acids by Self-assembled Sodium Deoxycholate.
pH-Dependent Chiral Recognition of D- and L-Arginine Derived Polyamidoamino Acids by Self-assembled Sodium Deoxycholate.
Polymers (2020-04-17)
Federica Lazzari, Bruce D Alexander, Robert M Dalgliesh, Jenny Alongi, Elisabetta Ranucci, Paolo Ferruti, Peter C Griffiths
ABSTRACT
D- and L-arginine-based polyamidoamino acids, called D- and L-ARGO7, retain the chirality and acid/base properties of the parent -amino acids and show pH-dependent self-structuring in water. The ability of the ARGO7 chiral isomers to selectively interact with chiral biomolecules and/or surfaces was studied by choosing sodium deoxycholate (NaDC) as a model chiral biomolecule for its ability to self-assembly into globular micelles, showing enantio-selectivity. To this purpose, mixtures of NaDC with D-, L- or D,L-ARGO7, respectively, in water were analysed by circular dichroism (CD) spectroscopy and small-angle neutron scattering (SANS) at different levels of acidity expressed in terms of pD and concentrations. Differences in the CD spectra indicated chiral discrimination for NaDC/ARGO7 mixtures in the gel phase (pD 7.30) but not in the solution phase (pD 9.06). SANS measurements confirmed large scale structural perturbation induced by this chiral discrimination in the gel phase yet no modulation of the structure in the solution phase. Together, these techniques shed light on the mechanism by which ARGO7 stereoisomers modify the morphology of NaDC micelles as a function of pH. This work demonstrates chirality-dependent interactions that drive structural evolution and phase behaviour of NaDC, opening the way for designing novel smart drug delivery systems.
Sigma-Aldrich
D-Arginine, ≥98% (TLC)
Pricing and availability is not currently available.