Merck
  • Systemic Exposure to Air Pollution Induces Oxidative Stress and Inflammation in Mouse Brain, Contributing to Neurodegeneration Onset.
Systemic Exposure to Air Pollution Induces Oxidative Stress and Inflammation in Mouse Brain, Contributing to Neurodegeneration Onset.
International journal of molecular sciences (2020-05-28)
Chiara Milani, Francesca Farina, Laura Botto, Luca Massimino, Elena Lonati, Elisabetta Donzelli, Elisa Ballarini, Luca Crippa, Paola Marmiroli, Alessandra Bulbarelli, Paola Palestini
ABSTRACT
In northern Italy, biomass burning-derived (BB) particles and diesel exhaust particles (DEP) are considered the most significant contributors to ultrafine particle (UFP) emission. However, a comparison between their impact on different brain regions was not investigated until now. Therefore, male BALB/c mice were treated with a single or three consecutive intratracheal instillations using 50 µg of UFPs in 100 µL of isotonic saline solution or 100 µL of isotonic saline solution alone, and brains were collected and analyzed. Proteins related to oxidative stress and inflammation, as well as Alzheimer's disease markers, were examined in the hippocampus, cerebellum, and the rest of the brain (RoB). Histopathological examination of the brain was also performed. Moreover, correlations among different brain, pulmonary, and cardiovascular markers were performed, allowing us to identify the potentially most stressful UFP source. Although both acute exposures induced inflammatory pathways in mouse brain, only DEP showed strong oxidative stress. The sub-acute exposure also induced the modulation of APP and BACE1 protein levels for both UFPs. We observed that DEP exposure is more harmful than BB, and this different response could be explained by this UFP's different chemical composition and reactivity.
Product Description
Supelco
Bicinchoninic Acid solution
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(II) sulfate solution, 4 % (w/v) (prepared from copper (II) sulfate pentahydrate)
Pricing and availability is not currently available.