- Experimental Analyses Emphasize the Stability of the Meisenheimer Complex in a SNAr Reaction Toward Trends in Reaction Pathways.
Experimental Analyses Emphasize the Stability of the Meisenheimer Complex in a SNAr Reaction Toward Trends in Reaction Pathways.
Frontiers in chemistry (2020-08-06)
Paola R Campodónico, Belén Olivares, Ricardo A Tapia
PMID32754575
ABSTRACT
The mechanism of SNAr reactions between 2-chloro-5-nitropyrimidine with primary and secondary alicyclic amines, respectively, have been studied by kinetic measurements. The kinetic data obtained in aqueous media opens a controversial discussion based on Brönsted-type plots analysis. The first approach based on the kinetic data reveals a non-catalyzed pathway. Then, the subtlety of the mathematical treatment of the kinetic data is discussed over a concerted or stepwise mechanism, respectively.