Merck
  • LncRNA SNHG16 Promotes the Progression of Laryngeal Squamous Cell Carcinoma by Mediating miR-877-5p/FOXP4 Axis.
LncRNA SNHG16 Promotes the Progression of Laryngeal Squamous Cell Carcinoma by Mediating miR-877-5p/FOXP4 Axis.
OncoTargets and therapy (2020-06-18)
Xiaoli Wang, Liming Liu, Wenfei Zhao, Qingyan Li, Guangsheng Wang, Huahui Li
ABSTRACT
Laryngeal cancer is a common malignant tumor in the ENT, of which laryngeal squamous cell carcinoma (LSCC) accounts for more than 90% of laryngeal cancer. The purpose of this study is to investigate the regulatory mechanism of lncRNA SNHG16 in LSCC. Real-time quantitative reverse transcription polymerase chain reaction (RT-qPCR) was used to measure mRNA expression. Cell Counting Kit (CCK-8), Transwell and luciferase reporter assays, flow cytometric analysis and Western blot analysis were used to investigate the function of lncRNA SNHG16 in LSCC. SNHG16 expression was increased in LSCC tissues and cells. The abnormal expression of SNHG16 was associated with clinical stage and lymph node metastasis in LSCC patients. In addition, knockdown of SNHG16 restrained cell proliferation, migration and invasion in LSCC. More importantly, SNHG16 acted as a competitive endogenous RNA in LSCC and regulated FOXP4 expression by making miR-877-5p sponge. Further, SNHG16 promoted LSCC progression by interacting with miR-877-5p and FOXP4. LncRNA SNHG16 promotes the progression of LSCC by sponging miR-877-5p and upregulating FOXP4.
Sigma-Aldrich
MISSION® esiRNA, targeting human FOXP4