Merck
  • BNIP-2 retards breast cancer cell migration by coupling microtubule-mediated GEF-H1 and RhoA activation.
BNIP-2 retards breast cancer cell migration by coupling microtubule-mediated GEF-H1 and RhoA activation.
Science advances (2020-08-14)
Meng Pan, Ti Weng Chew, Darren Chen Pei Wong, Jingwei Xiao, Hui Ting Ong, Jasmine Fei Li Chin, Boon Chuan Low
ABSTRACT
Microtubules display dynamic turnover during cell migration, leading to cell contractility and focal adhesion maturation regulated by Rho guanosine triphosphatase activity. This interplay between microtubules and actomyosin is mediated by guanine nucleotide exchange factor (GEF)-H1 released after microtubule depolymerization or microtubule disconnection from focal adhesions. However, how GEF-H1 activates Rho upon microtubule disassembly remains elusive. Here, we found that BNIP-2, a BCH domain-containing protein that binds both RhoA and GEF-H1 and traffics with kinesin-1 on microtubules, is important for GEF-H1-driven RhoA activation upon microtubule disassembly. Depletion of BNIP-2 in MDA-MB-231 breast cancer cells decreases RhoA activity and promotes cell migration. Upon nocodazole-induced microtubule disassembly, the interaction between BNIP-2 and GEF-H1 increases, while knockdown of BNIP-2 reduces RhoA activation and cell rounding via uncoupling RhoA-GEF-H1 interaction. Together, these findings revealed that BNIP-2 couples microtubules and focal adhesions via scaffolding GEF-H1 and RhoA, fine-tuning RhoA activity and cell migration.
Sigma-Aldrich
Anti-BNIP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human ARHGEF2
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Pricing and availability is not currently available.