Merck
  • β-Turn mimetic synthetic peptides as amyloid-β aggregation inhibitors.
β-Turn mimetic synthetic peptides as amyloid-β aggregation inhibitors.
Bioorganic chemistry (2020-07-20)
Stefanie Deike, Sven Rothemund, Bruno Voigt, Suman Samantray, Birgit Strodel, Wolfgang H Binder
ABSTRACT
Aggregation of amyloid peptides results in severe neurodegenerative diseases. While the fibril structures of Aβ40 and Aβ42 have been described recently, resolution of the aggregation pathway and evaluation of potent inhibitors still remains elusive, in particular in view of the hairpin-region of Aβ40. We here report the preparation of beta-turn mimetic conjugates containing synthetic turn mimetic structures in the turn region of Aβ40 and Aβ16-35, replacing 2 amino acids in the turn-region G25 - K28. The structure of the turn mimic induces both, acceleration of fibrillation and the complete inhibition of fibrillation, confirming the importance of the turn region on the aggregation. Replacing position G25-S26 provided the best inhibition effect for both beta-turn mimetics, the bicyclic BTD 1 and the aromatic TAA 2, while positions N27-K28 and V24-G25 showed only weaker or no inhibitory effects. When comparing different turn mimetics at the same position (G25-S26), conjugate 1a bearing the BTD turn showed the best inhibition of Aβ40 aggregation, while 5-amino-valeric acid 4a showed the weakest effect. Thus there is a pronounced impact on fibrillation with the chemical nature of the embedded beta-turn-mimic: the conformationally constrained turns 1 and 2 lead to a significantly reduced fibrillation, even inhibiting fibrillation of native Aβ40 when added in amounts down to 1/10, whereas the more flexible beta-turn-mimics 4-amino-benzoic acid 3a and 5-amino-valeric acid 4a lead to enhanced fibrillation. Toxicity-testing of the most successful conjugate showed only minor toxicity in cell-viability assays using the N2a cell line. Structural downsizing lead to the short fragment BTD/peptide Aβ16-35 as inhibitor of the aggregation of Aβ40, opening large potential for further small peptide based inhibitors.
Sigma-Aldrich
2-Iodoxybenzoic acid, contains stabilizer, 45 wt. % (IBX)
Pricing and availability is not currently available.