Merck
  • FoxO suppresses endoplasmic reticulum stress to inhibit growth of Tsc1-deficient tissues under nutrient restriction.
FoxO suppresses endoplasmic reticulum stress to inhibit growth of Tsc1-deficient tissues under nutrient restriction.
eLife (2020-06-12)
Avantika Gupta, Hugo Stocker
ABSTRACT
The transcription factor FoxO has been shown to block proliferation and progression in mTORC1-driven tumorigenesis but the picture of the relevant FoxO target genes remains incomplete. Here, we employed RNA-seq profiling on single clones isolated using laser capture microdissection from Drosophila larval eye imaginal discs to identify FoxO targets that restrict the proliferation of Tsc1-deficient cells under nutrient restriction (NR). Transcriptomics analysis revealed downregulation of endoplasmic reticulum-associated protein degradation pathway components upon foxo knockdown. Induction of ER stress pharmacologically or by suppression of other ER stress response pathway components led to an enhanced overgrowth of Tsc1 knockdown tissue. Increase of ER stress in Tsc1 loss-of-function cells upon foxo knockdown was also confirmed by elevated expression levels of known ER stress markers. These results highlight the role of FoxO in limiting ER stress to regulate Tsc1 mutant overgrowth.
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Pricing and availability is not currently available.
Sigma-Aldrich
Benzamidine, ≥95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Eeyarestatin I, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Pricing and availability is not currently available.