Merck
  • Region-Specific Transcriptional Control of Astrocyte Function Oversees Local Circuit Activities.
Region-Specific Transcriptional Control of Astrocyte Function Oversees Local Circuit Activities.
Neuron (2020-04-23)
Anna Yu-Szu Huang, Junsung Woo, Debosmita Sardar, Brittney Lozzi, Navish A Bosquez Huerta, Chia-Ching John Lin, Daniela Felice, Antrix Jain, Adriana Paulucci-Holthauzen, Benjamin Deneen
ABSTRACT
Astrocytes play essential roles in brain function by supporting synaptic connectivity and associated circuits. How these roles are regulated by transcription factors is unknown. Moreover, there is emerging evidence that astrocytes exhibit regional heterogeneity, and the mechanisms controlling this diversity remain nascent. Here, we show that conditional deletion of the transcription factor nuclear factor I-A (NFIA) in astrocytes in the adult brain results in region-specific alterations in morphology and physiology that are mediated by selective DNA binding. Disruptions in astrocyte function following loss of NFIA are most pronounced in the hippocampus, manifested by impaired interactions with neurons, coupled with diminution of learning and memory behaviors. These changes in hippocampal astrocytes did not affect basal neuronal properties but specifically inhibited synaptic plasticity, which is regulated by NFIA in astrocytes through calcium-dependent mechanisms. Together, our studies reveal region-specific transcriptional dependencies for astrocytes and identify astrocytic NFIA as a key transcriptional regulator of hippocampal circuits.
Sigma-Aldrich
Anti-NFIB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
D-Serine, ≥98% (TLC)
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Supelco
(R)-(+)-Limonene, analytical standard
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
MISSION® esiRNA, targeting human NFIB
Sigma-Aldrich
Anti-NFIA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
N-Acetyl-D-penicillamine, ≥99.0% (T)