Merck
  • Unravelling the anticancer potential of functionalized chromeno[2,3-b]pyridines for breast cancer treatment.
Unravelling the anticancer potential of functionalized chromeno[2,3-b]pyridines for breast cancer treatment.
Bioorganic chemistry (2020-05-26)
Sofia Oliveira-Pinto, Olívia Pontes, Diogo Lopes, Belém Sampaio-Marques, Marta D Costa, Luísa Carvalho, Céline S Gonçalves, Bruno M Costa, Patrícia Maciel, Paula Ludovico, Fátima Baltazar, Fernanda Proença, Marta Costa
ABSTRACT
A selection of new chromeno[2,3-b]pyridines was prepared from chromenylacrylonitriles and N-substituted piperazines, using a novel and efficient synthetic procedure. The compounds were tested for their anticancer activity using breast cancer cell lines MCF-7, Hs578t and MDA-MB-231 and the non-neoplastic cell line MCF-10A for toxicity evaluation. In general, compounds showed higher activity towards the luminal breast cancer subtype (MCF-7), competitive with the reference compound Doxorubicin. The in vivo toxicity assay using C. elegans demonstrated a safe profile for the most active compounds. Chromene 3f revealed a promising drug profile, inhibiting cell growth and proliferation, inducing cell cycle arrest in G2/M phase, apoptosis and microtubule destabilization. The new compounds presented exciting bioactive features and may be used as lead compounds in cancer related drug discovery.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
In Vitro Toxicology Assay Kit, Sulforhodamine B based