Merck
  • A novel structural class of potent inhibitors of NF-kappa B activation: structure-activity relationships and biological effects of 6-aminoquinazoline derivatives.
A novel structural class of potent inhibitors of NF-kappa B activation: structure-activity relationships and biological effects of 6-aminoquinazoline derivatives.
Bioorganic & medicinal chemistry (2003-08-21)
Masanori Tobe, Yoshiaki Isobe, Hideyuki Tomizawa, Takahiro Nagasaki, Hirotada Takahashi, Hideya Hayashi
ABSTRACT
In this study, we have investigated the roles of substituents on the terminal phenyl ring at the C(4)-position of the quinazoline core to complete the structure-activity relationships (SARs) of our NF-kappa B activation inhibitors. Among them, compound 12j afforded highly potent inhibitory activity toward NF-kappa B transcriptional activation with IC(50) value of 2 nM, along with an excellent in vivo efficacy by reducing the edema formation seen in carrageenin-induced inflammation of the rat hind paw.
Sigma-Aldrich
6-Aminoquinoline, 98%