Merck
  • Multiplexed Cre-dependent selection yields systemic AAVs for targeting distinct brain cell types.
Multiplexed Cre-dependent selection yields systemic AAVs for targeting distinct brain cell types.
Nature methods (2020-04-22)
Sripriya Ravindra Kumar, Timothy F Miles, Xinhong Chen, David Brown, Tatyana Dobreva, Qin Huang, Xiaozhe Ding, Yicheng Luo, Pétur H Einarsson, Alon Greenbaum, Min J Jang, Benjamin E Deverman, Viviana Gradinaru
ABSTRACT
Recombinant adeno-associated viruses (rAAVs) are efficient gene delivery vectors via intravenous delivery; however, natural serotypes display a finite set of tropisms. To expand their utility, we evolved AAV capsids to efficiently transduce specific cell types in adult mouse brains. Building upon our Cre-recombination-based AAV targeted evolution (CREATE) platform, we developed Multiplexed-CREATE (M-CREATE) to identify variants of interest in a given selection landscape through multiple positive and negative selection criteria. M-CREATE incorporates next-generation sequencing, synthetic library generation and a dedicated analysis pipeline. We have identified capsid variants that can transduce the central nervous system broadly, exhibit bias toward vascular cells and astrocytes, target neurons with greater specificity or cross the blood-brain barrier across diverse murine strains. Collectively, the M-CREATE methodology accelerates the discovery of capsids for use in neuroscience and gene-therapy applications.
Sigma-Aldrich
KIMBLE Dounce tissue grinder set, 2 mL complete
Supelco
4-(Methylnitrosoamino)-1-(3-pyridinyl)-1-butanone, analytical standard
Sigma-Aldrich
Anti-GLUT-1 Antibody, CT, from rabbit, purified by affinity chromatography