Merck
  • Hepatitis B virus X protein promotes liver cell pyroptosis under oxidative stress through NLRP3 inflammasome activation.
Hepatitis B virus X protein promotes liver cell pyroptosis under oxidative stress through NLRP3 inflammasome activation.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2020-04-30)
Wen-Hui Xie, Jian Ding, Xiao-Xia Xie, Xiao-Huang Yang, Xiao-Fan Wu, Zhi-Xin Chen, Qi-Lan Guo, Wen-Yu Gao, Xiao-Zhong Wang, Dan Li
ABSTRACT
Hepatitis B virus X protein (HBx) is a pivotal factor for HBV-induced hepatitis. Herein, we sought to investigate HBx-mediated NLR pyrin domain containing 3 (NLRP3) inflammasome activation and pyroptosis under oxidative stress. The effect of HBx on the NLRP3 inflammasome was analyzed by enzyme-linked immunosorbent assays, quantitative reverse transcription-polymerase chain reaction, western blotting, and immunofluorescence in hepatic HL7702 cells. Pyroptosis was evaluated by western blotting, lactate dehydrogenase release, propidium iodide staining, and transmission electron microscopy. NLRP3 expression in the inflammasome from liver tissues was assessed by immunohistochemistry. In hydrogen peroxide (H2O2)-stimulated HL7702 cells, HBx triggered the release of pro-inflammatory mediators apoptosis-associated speck-like protein containing a CARD (ASC), interleukin (IL)-1β, IL-18, and high-mobility group box 1 (HMGB1); activated NLRP3; and initiated pro-inflammatory cell death (pyroptosis). HBx localized to the mitochondria, where it induced mitochondrial damage and production of mitochondrial reactive oxygen species (mitoROS). Treatment of HL7702 cells with a mitoROS scavenger attenuated HBx-induced NLRP3 activation and pyroptosis. Expression levels of NLRP3, ASC, and IL-1β in liver tissues from patients were positively correlated with HBV DNA concentration. The NLRP3 inflammasome was activated by elevated mitoROS levels and mediated HBx-induced liver inflammation and hepatocellular pyroptosis under H2O2-stress conditions.
Sigma-Aldrich
Anti-Hepatitis B Virus Antibody, core Antigen (ayw), clone 10E11, a.a. 2-10, ascites fluid, clone 10E11, Chemicon®
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
MitoTEMPO, ≥98% (HPLC)