Merck
  • Cyclometallation of arylimines and nitrogen-containing heterocycles via room-temperature C-H bond activation with arene ruthenium(II) acetate complexes.
Cyclometallation of arylimines and nitrogen-containing heterocycles via room-temperature C-H bond activation with arene ruthenium(II) acetate complexes.
Dalton transactions (Cambridge, England : 2003) (2012-08-15)
Bin Li, Thierry Roisnel, Christophe Darcel, Pierre H Dixneuf
ABSTRACT
The reaction of [RuCl(2)(p-cymene)](2) with arylimines and 4 equiv. of KOAc in methanol at room temperature produces stable (N^C)-cyclometallated ruthenium(II) complexes via C-H bond activation/deprotonation. This method can be applied also to nitrogen-containing molecules: N-phenylpyrazole, 2-phenyl-2-oxazoline and benzo[h]quinoline. N-Phenyl-pyrazole, [RuCl(2)(p-cymene)](2) and diphenylacetylene directly lead to alkyne insertion into the metallacycle C-Ru bond.
Sigma-Aldrich
Chiralyst P612, Umicore, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Dichloro(p-cymene)ruthenium(II) dimer
Pricing and availability is not currently available.